Polypropylene Hinged Vials, Sterile, Qorpak®
Supplier: Qorpak
PLC-10051 PLC-10039 PLC-09774 PLC-10057 PLC-09773 PLC-10052 PLC-10045 PLC-10040 PLC-10056 PLC-09775
93001-672CS 280.1 CAD
93001-672 93001-678 93001-682 93001-688 93001-694 93001-696 93001-698 93001-700 93001-702 93001-716 93001-718 93001-720 93001-724 93001-730 89204-964
Polypropylene Hinged Vials, Sterile, Qorpak®
Vials
Lab grade polypropylene Hinged Vials are sterile, leak-proof and airtight.
The containers are manufactured in FDA approved Class 1000 and Class 100000 clean rooms. The containers are sterile by process and for most applications do not require a second means of sterilization. The containers are physically closed in the mold as part of the manufacturing process in clean rooms using Hepa-filtration systems, guaranteeing zero particulates larger than one half micron. They are non-absorptive and non-aersoling. The flip-top is integrally hinged to the container, allowing the user to open andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable