Polypropylene Vials, Starplex®
Supplier: Starplex Scientific
V112-YNL V8084-S V103-NL V102-NL V103-NL V8093 V102 V8095 V102-NL V8093-S V8094 V8084 V102-POUCH
CA73240-127CS 344.96 CAD
CA73240-127 76446-930 CA73240-129 14216-268 CA73240-133 CA10861-530 14216-270 CA11006-064 76417-694 CA73270-066 CA66410-024 76417-696 11007-280
Polypropylene Vials, Starplex®
Vials
These vials are made of crack-resistant medical grade polypropylene.
  • With or without screw caps
  • Flat, conical or round bottoms
Available clear for easy visual inspection and amber for light-sensitive components. These vials provide convenient storage for samples.