Nalgene® Sample Vials, Low-Density Polyethylene, with Snap Cap, Thermo Scientific
Supplier: Thermo Fisher Scientific
6250-0005 6250-0012 6250-0018 6250-9028 6250-9050 6250-9075
66018-109CS 301.35 CAD
66018-109 66018-142 66018-197 66018-212 66018-358 66018-359
Nalgene® Sample Vials, Low-Density Polyethylene, with Snap Cap, Thermo Scientific
Vials
With friction fit, snap closures.
Useful for samples, pH cups, weighing bottles, packing, and other general applications. Will not shatter like polystyrene vials.