Short Style Shell Vials, Borosilicate Glass, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
60965-4 74400-2040 60931-14 60931-12 60931-1 60931-2 60931-4 60931-6 60931-8
66012-285CS 508.05 CAD
66012-285 66023-007 66020-190 66020-191 66020-192 66020-194 66020-196 66020-197 66020-198
Short Style Shell Vials, Borosilicate Glass, Kimble Chase, DWK Life Sciences
Vials
Straight-sided shell vials are fabricated from clear N-51A borosilicate glass.
Ideal storage containers for dry products. Without caps.