Shorty Vials®, Borosilicate Glass, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
224821 224822 224823 224812 W225292 W225293 W224607 W225291 W224606 W224608
66011-868CS 240.86 CAD
66011-868 66011-879 66011-880 66011-915 80076-590 80076-592 80076-596 80076-588 80076-594 80076-598
Shorty Vials®, Borosilicate Glass, Wheaton, DWK Life Sciences
Vials
These shorty vials have a low profile for greater stability and reduced dead volume above the sample.
  • Made from low extractable borosilicate glass
  • Superior chemical resistance
Vials are available with or without caps and a choice of liners. PTFE faced rubber lined caps provide an inert and chemically-resistant barrier for harsh, sensitive, and reactive samples. Caps with 14B rubber liner are suitable for aqueous samples. Vials and caps are autoclavable.
Certifications: Vials meet ASTM E438 Type 1, Class A and USP Type 1 standards.
Ordering information: Vials are packaged in Lab File® partitioned, corrugated trays, then trays are shrink-wrapped to reduce particulate contamination.