Borosilicate Glass Vials in M-T Vial File® Storage Cases, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
224891 224892 224952 W224896 W224897 W224693 W224694 W224695 W224696 W224954 W224955
66011-653CS 183.69 CAD
66011-653 66011-675 66011-755 66014-506 80076-600 80076-602 80076-608 80076-610 80076-612 80076-614 80076-620 80076-622
Borosilicate Glass Vials in M-T Vial File® Storage Cases, Wheaton, DWK Life Sciences
Vials
Clear Wheaton 33® low extractable borosilicate glass vials and amber Wheaton 320 glass vials are packaged in a Vial File reusable transparent plastic case with alphanumeric grid.
Wheaton 33 glass protects pH balance and product integrity; amber glass provides protection for light-sensitive materials. Vials and caps are autoclavable. Aqueous sample vials have black phenolic solid-top caps with white 14B rubber liners. Organic sample vials have black phenolic fluoropolymer resin-faced liners with 14B rubber (solid-top caps) or silicone (open-top caps).
Certifications: Clear vials meetMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable