Ovation® Macro Single-Channel Electronic Pipettors, VistaLab
Supplier: VistaLab
1065-05ML 1065-10ML
89027-676EA 2021.5 CAD
89027-676 89049-038
Ovation® Macro Single-Channel Electronic Pipettors, VistaLab
Pipettes Micropipettes
The ideal device for repetitive pipetting of volumes over 1 ml, particularly in tight quarters.
  • High-precision alternative to using pipette controllers with serological pipettes
  • Excellent accuracy and precision
  • Fast, easy repeat dispensing
A relaxed grip design, low weight, and the ease of electronic operations facilitates repetitive pipetting applications.
Pipettors are fully electronic, with a broad pipetting range of 0.2 to 10 ml. Each pipettor allows rapid, accurate, and precise repeatable dispensing in addition to a host of other essential liquid handling functions. Setup and recall of volumes, speeds, repetitions, and other settings are easily determined by push-button functions. Tips simply click into place, and may be ejected at the push of a button, eliminating the inconveniences associated with traditional serological
pipetting.
A full line of standard, sterile, and sterile filtered tips in VistaClear™ recyclable packaging is also available. For occasions when extra length is needed for tall vessels, the device's unique nozzle can accommodate a serological pipet, and provide the same precise delivery as VistaClear™ pipettor tips.
Ordering information: All Ovation electronic pipettors are supplied with a rechargeable power supply.