PYREX® Solvent Still Head, Corning
Supplier: Corning
3575-250 3575-500
26307-220CS 1848.68 CAD
26307-220 26307-222
PYREX® Solvent Still Head, Corning
Distillation Components Distillation Heads
PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Solvent still head for maintaining oxygen-free solvents. Head has sufficient storage space to allow solvent removal by syringe or drainage through 2 mm T-bore fluoropolymer resin stopcock. It has a NS outer joint at top, NS inner joint at bottom, and NS 14/20 inner joint on the drainage arm.