Nalgene® Racks, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
5930-0016 5930-0020 5930-0030
60985-206CS 615.58 CAD
60985-206 60985-217 60985-228
Nalgene® Racks, Polypropylene, Thermo Scientific
Racks Tube Racks
Widely spaced holes and three-tiered design facilitate insertion and removal of tubes
Holes are keyed with molded numbers and letters for easy identification. Superior chemical resistance. Not autoclavable.
Order Now
SPECIFICATIONS
Environmentally Preferable