KONTES® Claisen Adapters for Distillation, Kimble Chase, DWK Life sciences
Supplier: DWK Life Sciences (KIMBLE)
161500-2440 273760-0000 273755-0000
KT161500-2440EA 285.15 CAD
KT161500-2440 KT161500-2942 KT273750-0000 KT273760-0000 KT273755-0000
KONTES® Claisen Adapters for Distillation, Kimble Chase, DWK Life sciences
Distillation Components Distillation Adapters
Provide dual entry into a variety of flasks.
  • Useful for reflux with addition-type reactions
  • Joints are the same size
Allow temperature measurement at either top outer opening when used in conjunction with a thermometer adapter. Useful for reflux with addition-type reactions.
Compatible with 179700 glass BEVEL-SEAL™ inlet adapters or 179800 PTFE BEVEL-SEAL™ inlet adapters for thermometer use at either top opening.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.