Nalgene® Micro Packaging Vials and Closures, PPCO, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
362800-0020 362805-0005 342800-0015 342800-0005 362805-0020 342810-0020 342805-0005 342800-0020 362800-0005 342805-0020 342810-0005 342800-0045
16210-876CS 319.06 CAD
16210-876 73521-160 16211-034 16210-872 16211-036 21020-106 16211-035 16210-870 21020-104 73521-158 16210-878 73520-780
Nalgene® Micro Packaging Vials and Closures, PPCO, Thermo Scientific
Vials
Single-use vials are ideal for air shipment of small-volume reagents and for many diagnostic kit applications.
High-purity, low metal-content polypropylene copolymer (PPCO) resin is ideal for PCR reagents. Vials can be centrifuged to 13000 g at room temperature. Precise external dimensions for smooth, jam-free automated filling. Leakproof, threaded screw closures have no O-ring to fall out or contaminate contents. Vials and closures are noncytotoxic and nonpyrogenic. The 11 mm closures fit the 0.5, 1.5, and 2 ml vials. The 13 mm closures fit the 4.5 ml vials. Vials 73521-158 and 73521-160 are sterilized using electron beam irradiation.
Certifications: Vials and closures meet FDA CFR21 177.1520 and USP Class VI requirements. Amber vials and closures meet USP XXIV 661 requirements for light transmission.
Ordering information: Nonsterile closures are bulk packed. They are sold separately and are available on the Accessories tab.