Eppendorf twin.tec® Real-Time PCR Plates, 96-Well, qPCR Plates
Supplier: Eppendorf
951022015 951022055 0030132718 951022003 951022043 0030132700 0030129644 0030129636
CA89087-438CS 328.36 CAD
CA89087-438 CA89087-444 CA89136-716 CA89087-436 CA89087-442 CA89136-714 76019-000 76019-002
Eppendorf twin.tec® Real-Time PCR Plates, 96-Well, qPCR Plates
Microplates PCR Plates
Eppendorf twin.tec® real-time PCR plates, designed for low-volume qPCR, utilize white wells to enhance fluorescent light reflection, improving sensitivity and reproducibility in assays for reliable results.
  • White wells for better reflection
  • High mechanical stability
  • Effective sealing by raised well rims
  • Skirted and semi-skirted frames
  • Optimal heat transfer due to reduced wall thickness
  • Autoclavable (121 °C, 20 min)
  • Available in lot-certified PCR clean and Eppendorf Forensic DNA Grade for tested purity
The limiting factor in low-volume real-time PCR (qPCR) is often the remaining fluorescence intensity. Eppendorf twin.tec real-time PCR plates are specially manufactured for qPCR, as they offer all the advantages of regular twin.tec plates, but with the added advantage of white wells. These reflect fluorescence much better than clear or frosted wells, so lower levels of fluorescence are still detectable with the same instrument - just by using the right consumables. The qPCR plates white wells also significantly reduce interfering background fluorescence and lead to increased consistency between replicates, and giving highly reproducible results.
The wells are made from polypropylene - which guarantees fast heat transfer for fast assays - and the frame is made of rigid polycarbonate, to ensure easy and reliable handling - both in manual and automated assays.
Autoclavable: 121 °C, 20 min.
Centrifugation stability: 6000×g