Nalgene® Unwire™ Racks, ResMer® Resin, Thermo Scientific
Supplier: Thermo Fisher Scientific
5970-0013 5970-0113 5970-0213 5970-0313 5970-0413 5970-0513 5970-0016 5970-0116 5970-0216 5970-0316 5970-0416 5970-0516 5970-0020 5970-0120 5970-0220 5970-0320 5970-0420 5970-0520 5970-0025 5970-0125 5970-0225 5970-0325 5970-0425 5970-0525 5970-0030 5970-0130 5970-0230 5970-0330 5970-0430 5970-0530
60914-605CS 479.55 CAD
60914-605 60914-616 60914-627 60914-638 60914-649 60914-660 60914-707 60914-718 60914-729 60914-730 60914-740 60914-762 60914-809 60914-820 60914-842 60914-864 60914-875 60914-886 60914-900 60914-922 60914-944 60914-966 60914-977 60914-988 60915-030 60915-051 60915-073 60915-095 60915-110 60915-131
Nalgene® Unwire™ Racks, ResMer® Resin, Thermo Scientific
Racks Tube Racks
These durable, brightly colored racks feature a three-tier design
Racks for 13mm and 16mm tubes hold 72 tubes; racks for 20mm and 25mm tubes hold 40 tubes; racks for 30mm tubes holds 24 tubes. Autoclavable.
They have excellent chemical resistance and can be immersed in a water bath. Letters and numbers on top tier aid in tube identification. Labels can be placed on large end plates. ResMer® technology allows these racks to be used over a wide temperature range.