SP Bel-Art Poxygrid® Centrifuge Tube Racks, 50 ml, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18794-0000 F18794-0001 F18794-0002
21008-485CS 1401.52 CAD
21008-485 21008-486 21008-487
SP Bel-Art Poxygrid® Centrifuge Tube Racks, 50 ml, Bel-Art Products, a part of SP
Racks Tube Racks
Made of epoxy-coated steel wire, these racks hold up to thirty-six 50 mL centrifuge tubes in holes 30 mm square.
  • Epoxy-coated steel construction
  • Inert
  • Autoclavable
  • Hold Petri dishes securely
Heat-cured epoxy coating is unaffected by organic solvents, salts, weak acids, and weak alkalis
These poxygrid® 50 mL centrifuge tube racks are made of plastic.
Racks will withstand a temperature range of -223ºC (-369ºF) to 121ºC (250ºF) and can be autoclaved.