Distillation Apparatus, Still Body, Short Path, Ace Glass
Supplier: Ace Glass
9317-11 9317-17 9317-42 9317-50 9317-52 9317-60 9317-03 9317-21 9317-30 9317-35
89074-366EA 545.63 CAD
89074-366 89074-368 89074-372 89074-374 89074-376 89074-378 89074-380 89074-382 89085-088 89085-090
Distillation Apparatus, Still Body, Short Path, Ace Glass
Distillation Components Distillation Sets
This short-path distillation apparatus is designed for small quantities, where holdup between flask and receiver must be minimized, but where it is also desired to measure vapor temperature with the accuracy of larger apparatus
The still body is available with or without a vacuum jacket, and with or without vigreux indents (which provide greater efficiency). The upper hose connections are for water lines; the lower connection for vacuum or vent. The main joints on the apparatus are [ST] 14/20. Thermometer joint and bleed tube are [ST] 10/18. Cow receiver is one piece. Hose connections are size A, for use with 5/16" I.D. tubing. Apparatus is fabricated from borosilicate glass.
Ordering information: Available complete in jacketed, unjacketed, and jacketed with indents versions, or components can be purchased separately. Complete items include [ST] 14/20-10/18 still body, [ST] 14/20 cow receiver, [ST] 10/18 bleed tube, and [ST] 14/20-10/18 flask.