KIMAX® Apparatus with Graham Condenser, [ST] Joint, and [ST] Stopper, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
21500-500 21500-1000
89001-740CS 1066.72 CAD
89001-740 89001-738
KIMAX® Apparatus with Graham Condenser, [ST] Joint, and [ST] Stopper, Kimble Chase
Distillation Components Distillation Sets
Apparatus used in general purpose distillation and phenol distillation
Flask has a flat bottom and a distillation head with a [ST] 19/38 joint. Supplied with a [ST] stopper. Condenser is Graham-style with a 200mm jacket that has a [ST] 19/38 joint at the top.