Synthware Distillation Apparatus, Microscale, Kemtech America
Supplier: Kemtech America
D151814 D151819
60002-056EA 369.38 CAD
60002-056 60002-058
Synthware Distillation Apparatus, Microscale, Kemtech America
Distillation Components Distillation Sets
Short-path design for microscale applications
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Jacketed
  • Microscale
  • Drip tip
  • Designated 10/18 thermometer joint
Used with a distillation receiver and 25mm immersion thermometer. Thermometer [ST] joint size: 10/18.
Ordering information: Distillation receiver and thermometer are not supplied. For distallation receiver, see 60002-060 series. For thermometer, contact your Avantor representative.