Nalgene® Cryogenic Vial Closure Color Coders, Polystyrene, Thermo Scientific
Supplier: Thermo Fisher Scientific
5045-0000 5045-0002 5045-0003 5045-0004 5045-0005
24270-056CS 179.58 CAD
24270-056 24270-058 24270-060 24270-062 24270-064
Nalgene® Cryogenic Vial Closure Color Coders, Polystyrene, Thermo Scientific
Cap Inserts
Colorful plastic disks fit into top of NALGENE® cryogenic vial closure (1.2, 2.0 or 5.0mL vial).
Provide quick visual identification of specific vials to simplify inventory maintenance. Remain firmly in place during storage in mechanical and liquid-nitrogen freezers. Disks have cut-out top to facilitate handling of vials with forceps. Will not interfere with closure removal, vial handling or storage in boxes or canes. Flat surface permits writing on top of vial.