VWR® Sharps Container Systems, Multiple Colors and Styles
Supplier: VWR
VWR®
19001-006 19001-049 19001-041 19001-047 19001-043 19001-028 8708TV 19001-008 19001-001 19001-016 19001-023 19001-045 19001-030 19001-005 19001-020 19001-039 19001-010 19001-026 19001-003 19001-013 19001-025
19001-008CS 318.24 CAD
19001-008 19001-006 19001-028 19001-039 19001-049 19001-016 19001-005 19001-026 75800-272 19001-047 19001-003 19001-025 19001-013 19001-001 19001-023 19001-045 19001-043 19001-010 19001-020 19001-041 19001-030
VWR® Sharps Container Systems, Multiple Colors and Styles
Bins Sharps Containers
Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes, and other sharps.
  • Puncture-resistant
  • Autoclave-safe and incinerator-safe
  • Transparent, locking lids
  • Needle removal ports
The stackable sharps containers are puncture-resistant and both autoclaves-safe and incinerator-safe. The tortuous path systems restrict hand entry. The extended neck sharps containers feature a funneled vertical drop and extended handles.