SP Bel-Art Poxygrid® Glove Dispenser Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F24739-0002 F24739-0003 F24739-0001
74950-160EA 142.18 CAD
74950-160 74950-162 74950-158
SP Bel-Art Poxygrid® Glove Dispenser Racks, Bel-Art Products, a part of SP
Glove Dispensers and Glove Box Holders
Durable, blue epoxy-coated welded steel wire racks keep disposable gloves or wipes handy.
  • Epoxy-coated welded steel wire construction
  • Durable
  • Autoclavable
  • Dispensers securely hold disposable gloves
  • Mounted to a vertical surface
Dispensers hold one, two, or three boxes up to 12.7Wx9.5Dcm (5x33/4") in size. Designed to be mounted to a vertical surface. Autoclavable.
Order Now
SPECIFICATIONS
Environmentally Preferable