Flip-Top Finger Cot Dispenser, AK®
Supplier: AK
AK-901
32799-056EA 241.53 CAD
32799-056
Flip-Top Finger Cot Dispenser, AK®
Glove Dispensers and Glove Box Holders
Cleanroom finger cot dispenser features hinged lid and door for easy loading and removal.
  • Made of clear 5 mm (3/16") acrylic
  • Openings feature radius edges for a smooth finish
Dispenser can be placed on a countertop or mounted on a wall.