Somso® Model of the Head
Supplier: MARCUS SOMMER
BS8
470313-140EA 1589.6 CAD
470313-140
Somso® Model of the Head
Models Human Biology Models
Somso® premium quality models.
  • 5 year warranty
Natural size, in SOMSO-Plast®. Showing, in right median section, the muscles, nerves and vessels in particular the cavities of nose and mouth. In one piece. Mounted on a green board under a removable transparent cover.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.