Pliers Decappers, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
224372 224373
26676-428CS 285.04 CAD
26676-428 74830-118
Pliers Decappers, Wheaton
Crimpers, Cappers and Decappers
Pliers decappers are an economical and easy way to remove seals.
To remove a seal, simply secure the seal between the jaws, squeeze the handles and twist the seal. Constructed of steel with red plastic-coated handles.
Order Now
SPECIFICATIONS
Environmentally Preferable