WHEATON® E-Z Decappers™ Hand-Operated Decappers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W225350 W225351 W225352 W225353
80078-998EA 1475.38 CAD
80078-998 80079-000 80079-002 80079-004
WHEATON® E-Z Decappers™ Hand-Operated Decappers, DWK Life Sciences
Crimpers, Cappers and Decappers
These durable, hand-operated decappers are an economical way to remove seals quickly and easily.
Note: Autoclaving may change the appearance or reduce product life.
To remove a seal, just place the decapper on the seal and squeeze the handles together. Decappers feature a cushioned ergonomic handle that helps reduce hand fatigue and adds comfort. The 8, 11, and 13mm models feature tapered jaws for easier vial access in racks. Each decapper size is labeled for quick identification. Autoclavable at 121°C (250°F).
Ordering information: Decappers work with the Crimpmaster® crimping station.