Vial Crushers, Thermo Scientific
Supplier: Thermo Scientific
C4040-VC
66064-904EA 19113.89 CAD
66064-904
Vial Crushers, Thermo Scientific
Mills
Unit breaks and separates liquid from vials and ampules for environmentally responsible waste handling.
  • Reduces overall waste disposal costs by reducing the total waste volume
  • Accomodates vials from 1 ml to 100 ml
  • Can handle approximately 5000×2 ml vials per hour
Safe to use in a fume hood or to connect to a vacuum system to limit exposure to flammable or toxic vapors. Crushed glass is automatically separated from solvent for separate disposal.