8-425 Screw-Thread Vials, Standard Opening, SUN-SRi
Supplier: SUN SRi
200 252 200 250
66008-042PK 129.7 CAD
66008-042 66008-264
8-425 Screw-Thread Vials, Standard Opening, SUN-SRi
Vials
Vials are manufactured of clear, Type I Class A or amber, Type I Class B borosilicate glass.
  • Standard graduated write-on patch for easy filling and identification
Vials require the use of a 8-425 closure and 8 mm septa. Preassembled caps and septa kits are available separately. These kits reduce the chance of contamination of the septum during sample preparation and save sample preparation time.
Delivery information: Kits feature polypropylene caps and PTFE/silicone septa.