VWR® Disposable Aluminum Weighing and Drying Pans
Supplier: VWR International
D-125 D-11 D-12 D-126-50 D-123
12175-001CS 705.22 CAD
12175-001 12177-266 12177-299 12177-301 12177-305
VWR® Disposable Aluminum Weighing and Drying Pans
Boats Weighing Boats
These oil-free pans are ideal for general-purpose weighing, dispensing, storing, and drying applications.
  • Sturdy construction
  • Compatible with most balances and dryers
  • Ideal for general-purpose applications
  • Disposable
  • Aluminum construction
Sturdy, with a slight taper for easy stacking. The 7 cm (23/4") I.D. pan is ideal for enclosed balances and weighing equipment with a 7 to 9.5 cm (23/4 to 33/4") diameter head.