Testing Jars, Clear, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
5910243C-21 5910243C-26 5910243B
89046-962CS 1071.04 CAD
89046-962 89046-964 89046-966
Testing Jars, Clear, Kimble Chase, DWK Life Sciences
Jars General Use Jars
Jars feature a wide-mouth design for efficient addition and removal contents
Clear glass allows for easy viewing of contents. Made of Type III soda-lime glass. Packaged in bulk packs (shrink modules with caps in bags).
Order Now
SPECIFICATIONS
Environmentally Preferable