Bouffant Cap Dispenser, S-Curve Technologies
Supplier: S-Curve Technologies
BD-123 PETG BD-600 PETG BD-600 BD-800PETG BD-808PETG
10693-138EA 295.67 CAD
10693-138 98106-944 89208-666 10693-140 32948-920
Bouffant Cap Dispenser, S-Curve Technologies
PPE Dispensers
Bouffant cap dispensers are designed to provide safe and convenient storage and access to large volumes of caps, beard covers, or similarly packaged garments.
  • Dispensers can be wall-mounted or used free-standing
  • Edges are flame-treated for smoothness
All dispensers are equipped with a full-length hinged lid for quick and easy interior access. Available in either acrylic or PETG construction, all units have a thickness of 3 mm (1/8").