Combs for Owl™ Large Gel System, Models A1, A2 and A3, Thermo Scientific
Supplier: Thermo Scientific
Gator™ Buffer Puffer™
A2-MT2C A2-MT2D A2-36C A2-8D A2-32C A2-12C A2-16D A1-PREP A1-RL-25D A1-12C A2-28D A2-PREP A1-RL-9C A2-16C A1-MTD A2-MTD A2-12D A1-24D A2-RL-18D A2-32D A2-36D A1-8D A1-8C A2-20C A1-24C A1-MTC A1-16C A1-16D A1-RL-9D A2-20D A2-28C A1-20D A1-WALL A2-24C A2-MTC A2-8C A1-12D A1-20C A2-24D
27372-044EA 123.75 CAD
27372-044 27372-088 27372-066 27372-100 27372-068 27372-102 27372-062 27372-086 27372-064 27372-120 27372-080 27373-302 27373-300 27372-060 27373-306 27373-260 27373-288 27373-286 27372-108 27372-048 27372-104 27372-106 27372-110 27372-078 27372-056 27372-079 27372-058 27372-096 27372-052 27372-054 27372-098 27372-076 27372-092 27372-050 27372-094 27372-090 27372-118 27373-290 27372-116
Combs for Owl™ Large Gel System, Models A1, A2 and A3, Thermo Scientific
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Combs are 1.0 mm or 1.5 mm thick
Microtiter combs with 21 and 42 teeth are spaced for microtitration of every other well.