Apparel Dispensers, S-Curve Technologies
Supplier: S-Curve Technologies
AD-101 AD-101 PETG
14007-126CS 1109.59 CAD
14007-126 89208-658 89208-660
Apparel Dispensers, S-Curve Technologies
PPE Dispensers
One-compartment apparel dispensers are designed to dispense aprons, coveralls, frocks, and smocks
They feature an open front for easy access and are made of durable clear acrylic or PETG with a thickness of 6 mm (¹/₄"). Dispensers can be mounted to the wall or used free-standing. Edges are flame-treated for smoothness. Available with or without a lid.
Ordering information: Supplied with heavy-duty mounting bracket and support bar. Dimensions: 61 x 38.5 x 26 cm (24 x 15.5 x 10.5")