Combs for Owl™ Multiple Gradient Gel Caster, Models P1 and P2, Thermo Scientific
Supplier: Thermo Fisher Scientific
P1-10C P1-10D P1-15C P1-15D P1-24C P1-24D P2-PREP P2-10C P2-10D P2-15C P2-15D P2-20C P2-25C
27372-512EA 110.1 CAD
27372-512 27372-514 27372-526 27372-528 27372-530 27372-532 27372-534 27372-536 27372-558 27372-566 27372-568 27372-570 27372-572 27372-576 27372-588
Combs for Owl™ Multiple Gradient Gel Caster, Models P1 and P2, Thermo Scientific
Electrophoresis Systems Vertical Gel Electrophoresis Systems
Choose from standard, preparative, well/sequencing, sharktooth, and sharktooth microtiter format combs.
Sharktooth combs with 36 teeth are spaced for microtitration of every other well. Sharktooth combs with 55 and 110 teeth are spaced for microtitration of every third well. Sharktooth combs with 73 and 146 teeth are spaced for microtitration of every fourth well.