BRAND® seripettor® pro Bottle-Top Dispenser, BrandTech
Supplier: Brandtech
4720450 4720420 4720440
89042-166EA 1795.09 CAD
89042-166 89136-056 89042-164
BRAND® seripettor® pro Bottle-Top Dispenser, BrandTech
Bottle Top Dispensers
Designed to handle a wider range of liquids than the standard seripettor®, the seripettor pro features an easily replaceable wiping seal dispensing cartridge, making it particularly suitable for use with crystal-forming liquids
  • Enhanced compatibility compared to seripettor®
  • Replaceable dispensing cartridge for long service life
  • Includes adapters to fit 45 mm, 40 mm, 38 mm and 33 mm bottle threads
Note: Valves and seals on the seripettor® pro allow for a broad range of chemicals and liquids to be dispensed. Chemicals incompatible with the materials of construction (HDPE, PP, borosilicate glass, platinum iridium, PTFE, ETFE, FEP, Al2O3 ceramic) must never be used with the seipettor® pro. These unsuitable chemicals include, but are not limited to: carbon disulfide; hydrofluoric acid and sodium azide solutions.
The replaceable cartridge facilitates quick and simple service, providing a quick-change option, and minimum downtime in the event of damage. The pump assembly is fitted with a corrosion-resistant stainless steel spring, for semi-automatic, one-handed dispensing. An opaque upper sleeve protects light-sensitive liquids while permitting cylinder observation to confirm the purging of bubbles during dispensing. The telescoping filling tube adjusts to fit a wide array of bottle sizes without cutting. Threaded safety cap provides a secure seal, and unscrews easily with one hand.
Temperature range: 15 to 40°C (59 to 104°F)
Ordering information: Each seripettor pro dispenser is supplied with a one-year manufacturer's warranty, spare dispensing cartridge, flexible discharge tube, telescoping filling tube, two polypropylene adapters (GL45/33 and GL45/38), and operating manual.