SP Bel-Art Lab-Aire® II Electric Benchtop Glassware Drying Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18819-0012 F18819-0023 F18819-0013
89050-336EA 2041.76 CAD
89050-336 89050-360 89050-362 89050-364
SP Bel-Art Lab-Aire® II Electric Benchtop Glassware Drying Racks, Bel-Art Products, a part of SP
Racks Draining Racks
Lab-Aire® II Electric Drying Racks accelerate drying of labware with warm, circulating air while their compact footprint uses minimal benchtop space.
  • Rugged ABS plastic construction
  • Easy to clean
  • Corrosion-resistant
  • Removable baskets serve as storage for small labware
  • Sturdy bases for excellent balance
400 watts of drying power gently circulates heated air into and around items to decrease drying time and efficiency is enhanced with a replaceable foam filter that keeps warmed air dust free
Made of rugged ABS plastic, they will never corrode and easily wipe clean. Drip/storage trays have scalloped wells to hold long thin labware. Small (3"), medium (4") and large (5") pegs can be moved and repositioned to hold a variety of labware including beakers, flasks, bottles, test tubes and cylinders. To redirect air flow, simply insert supplied plugs instead of pegs into unused peg holes. Removable polypropylene baskets serve as storage for small labware and extra pegs.
The heat selector knob allows the choice of three temperature settings: high, medium and low. The power button illuminates when in use, and an 8-amp @ 120V circuit breaker and independent thermo-fuse protects the unit from overheating. Freestanding with sturdy bases and excellent balance, they can be placed on any flat surface.
Certifications: CE approved.
Ordering information: Choose a single sided model to fit nicely against a wall for economy of space, or double the capacity with a minimal increase in footprint with a double-sided model. Extra accessory pegs are available to create a fully customizable drying system. Pegs measuring 3" (89050-352), 4" (89050-354), and 5" (89050-356) are available separately. Additional baskets are also available (see 60914-208 series).