VWR® Talon® Support Stand, Cast Iron, U-Shaped Base
Supplier: VWR
VWR® Talon®
915385
12985-078EA 109.1 CAD
12985-078
VWR® Talon® Support Stand, Cast Iron, U-Shaped Base
Retort Stands
Support stand base is constructed of cast iron with black enamel finish.
  • Cast iron construction with black enamel
  • U-shaped base
  • Built-in support rod holder
  • For use with VWR® Talon® lab frames
Base features three built-in support rod holders for additional support or to customize your stand. Locking knob accepts a 1.3 cm (1/2") diameter support rod (sold separately).