Pre-Cleaned Wide-Mouth Glass Jars, Clear, Environmental Express®
Supplier: Environmental Express
APC3110 APC3050 APC1070 APC1110 APC3989
23050-038CS 222.23 CAD
23050-038 23050-140 23050-144 23050-334 89202-792 89202-794 89202-852
Pre-Cleaned Wide-Mouth Glass Jars, Clear, Environmental Express®
Jars General Use Jars
All containers meet one of two levels of pre-cleaning.
Amber glass jars include polypropylene closure and PTFE liner.
Level III indicates that containers have not received an EPA washing treatment. Level I indicates that each case of containers has been custody sealed, lot number labeled for traceability, and a Certificate of Analysis for EPA quality assurance treatment.
Order Now
SPECIFICATIONS
Environmentally Preferable