Shell Vials
Supplier: Thermo Scientific
C4015-48 C4008-50 C4015-96PA C4011-77P C4015-94 C4015-95P 1-NWV-I
66011-204PK 184.89 CAD
66011-204 66011-206 66011-212 66030-362 66030-834 66030-940 66064-894 66065-386 66065-390 66065-392 66065-666 66065-668 66064-326 10017-810
Shell Vials
Vials
The 1 ml, 8×35 mm vials are designed for use in Alcott model 728 autosamplers. The 1 ml, 8×40 mm vials are specifically designed for Waters® 96-position trays. The 2 ml vial will work on any standard size 2 ml tray. The 4 ml vial is compatible with Waters® 48-position trays. Shell vials are manufactured from borosilicate glass unless otherwise specified.
  • Molded sealing ring secures sample containment
SepCap™ closures are designed for easy syringe needle penetration and easy push-in/pull-out from vial.
Conical-style Polyspring® inserts are available in borosilicate glass or polypropylene.
Certifications: Certified vial kits are tested and certified to meet or exceed up to 17 critical performance parameters; certificate enclosed with kit.
Delivery information: Vials are supplied with polyethylene push-in caps or polypropylene SepCap™ closures. Convenience kits include vials and caps with pre-inserted septa.