Optifix® Adapters, MilliporeSigma
Supplier: MilliporeSigma
109038-1 109058-1 109025-1 109028-1 109030-1 109036-1 109050-1 109052-1 109065-1 109125-1 109128-1 109136-1 109138-1 109219-1
CA109038-1EA 121 CAD
CA109038-1 CAE19045-01 CA109058-1 CA109129-1 CAE18059-01 CAE19025-01 CAE19028-01 CAE19030-01 CAE19036-01 CAE19040-01 CAE19050-01 CAE19052-01 CAE19065-01 CAE19124-01 CAE19125-01 CAE19128-01 CAE19130-01 CAE19136-01 CAE19138-01 CAE19140-01 CAE19219-01 CAE19229-01 CAE19245-01
Optifix® Adapters, MilliporeSigma
Bottle Top Dispensers
For use with Optifix® and POLYFIX® dispensers (see EM-10108020-1 and EM-10107038-1 series)
Adapters fit standard screw-neck reagent bottles, and containers up to 200L drums. Sizes based on O.D. measurements.
Ordering information: Adapter set (EM-109050-1) includes four adapters: 24mm fits 4oz., 500mL, and 32oz. bottles; 28mm fits 4oz., 500mL, 16oz., 32oz., and 1L bottles; 38mm fits 1gal., 2.5L, 4L, 5gal., and 10gal. containers; and 45mm fits 10L containers. For additional adapter sizes, contact your Avantor sales representative.
Order Now
SPECIFICATIONS
Environmentally Preferable