Optifix® Dispensers, MilliporeSigma
Supplier: MilliporeSigma
10108044-1 10108048-1 10108058-1 10108133-1 10108144-1 10108153-1 10110638-1 10110644-1 108056G-1 10110738-1 10110748-1 10108027-1 10108033-1 10108051-1 10108127-1 10108138-1 10108148-1 108056-1 108053-1 10110727-1
CA10108044-1EA 2485 CAD
CA10108044-1 CA10108048-1 CA10108058-1 CA10108133-1 CA10108144-1 CA10108153-1 CA10110638-1 CA10110644-1 CA108056G-1 CAM10110738-1 CAM10110748-1 CA10108027-1 CA10108033-1 CA10108038-1 CA10108051-1 CA10108127-1 CA10108138-1 CA10108148-1 CA108056-01 CAE18053-01 CAM10110727-1
Optifix® Dispensers, MilliporeSigma
Bottle Top Dispensers
Optifix® Dispensers are available in four models, each of which is designed to increase efficiency and provide additional safety to daily laboratory operations. These dispensers are manufactured to the highest quality standards, ensuring accurate, reliable, and easy dispensing.
Ideal for critical dispensing applications, the Optifix® BASIC dispenser is used for aqueous solutions, light acids, and crystallizing substances
The Optifix® SOLVENT dispenser is useful for all organic solvents and materials that swell PTFE. The piston is glass. The Optifix® SOLVENT dispenser with vapor trap is designed specifically to trap organic vapors, moisture, and/or gases before they enter the bottle and contaminate the solvent.
The Optifix® SAFETY dispenser is for strong fuming acids, brominated compounds, and strong-smelling reagents. The piston is glass sheathed in PTFE and made with an additional lipseal to prevent fumes from escaping.
The Optifix® HF dispenser is for hydrofluoric acid. The dispenser construction is almost all PTFE.
The piston is glass sheathed in thick PTFE.
Ordering information: Complete with intake tube and Certificate of Precision. Bottles are not included. BASIC and SOLVENT models also include a set of three polypropylene adapters (38mm, 40mm, and 45mm). For the SAFETY and HF models, a PTFE adapter is recommended (see EM-109050-1 series).