VWR® Talon® Hook Connectors
Supplier: VWR International
12985-062 60079-166
12985-062EA 60.72 CAD
12985-062 60079-166
VWR® Talon® Hook Connectors
Retort Rods
Hook connectors allow one-handed assembly of two components with one adjustment screw.
  • Simple
  • Versatile
  • Easy to use
Maximum grip size: 13 mm (1/2").