700 System Premium Pistons, Techcon
Supplier: Techcon Systems
7100009WPK 7030006RPK 7030009WPK 7050006RPK 7100006RPK 7300006RPK 7300009WPK 7050009WPK
14219-634PK 101.7 CAD
14219-634 14219-640 14219-642 14219-654 14219-668 14219-680 14219-682 14219-710
700 System Premium Pistons, Techcon
Fluid Adhesive Dispensers and Pistons
Pistons are available in white wiper and red straight wall styles
The low friction wiper piston can be used with all materials and features a full lip seal. The air-free loading design of the straight wall piston allows air to bleed while retaining product security. Made of polyethylene. UV blocking. Dispense high-viscosity materials of 55,000cP minimum.