VWR® Talon® Horizontal Mounting Bars with Coupler
Supplier: VWR International
60079-104 60079-106 60079-107
60079-104EA 9.04 CAD
60079-104 60079-106 60079-107
VWR® Talon® Horizontal Mounting Bars with Coupler
Retort Rods
Horizontal mounting bars feature a threaded end for direct installation into channels.
  • For use with steel channel frames
  • Locking nut included
  • Provided with channel coupler
Supplied with spring coupler for use in 41mm (119/32") steel channels.