VWR® Talon® Rods, Stainless Steel
Supplier: VWR International
60079-530 60079-531 60079-532 60079-533 60079-534 60079-535 60079-536 60079-537 60079-538 60079-539 60079-540
60079-530CS 179.36 CAD
60079-530 60079-531 60079-532 60079-533 60079-534 60079-535 60079-536 60079-537 60079-538 60079-539 60079-540
VWR® Talon® Rods, Stainless Steel
Retort Rods
Choose VWR® top-of-the-line rods when you need maximum strength and durability.
  • Strong and resilient
  • For use with VWR® Talon® lab frames
  • Stainless steel construction
Constructed of Type 303 stainless steel.