SP Bel-Art Mason Jars, Wide Mouth, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art
F10916-0000 F10914-0000 F10913-0000 F10917-0000 F10915-0000
16128-660CS 606.91 CAD
16128-660 16128-626 16128-604 16128-648 70690-010
SP Bel-Art Mason Jars, Wide Mouth, Polypropylene, Bel-Art Products, a part of SP
Jars General Use Jars
Highly shock-resistant polypropylene, heavy-walled Mason Jars are ideal for industrial waste sampling, long-term storage, and a variety of laboratory uses.
  • Polypropylene construction
  • Shock-resistant
  • Heavy-walled design
  • Autoclavable
They have standard 70 mm, G-threaded necks, and screw caps with unique grip lugs and a removable leakproof silicone rubber ring seal.
Cap and jar are steam autoclavable at 121°C (250°F). Loosen cap when autoclaving.