PYREX® Cylindrical Jars, Corning
Supplier: Corning
6942-9L 6942-17L 6942-27L 6942-32L
36360-143CS 1499.48 CAD
36360-143 36360-187 36360-201 36360-245
PYREX® Cylindrical Jars, Corning
Jars General Use Jars
These PYREX® jars are mold-blown with substantial walls for mechanical strength.
  • Open end is ground flat
These jars can also be used as animal jars.
Caution: Do not use with heat, vacuum, or pressure applications.