PYREX® Cylindrical Chromatography Jars, Corning
Supplier: Corning
6945-13L 6945-6L 6945-36L
89091-152CS 6138.77 CAD
89091-152 36360-289 36360-369
PYREX® Cylindrical Chromatography Jars, Corning
Jars General Use Jars
These general purpose cylindrical chromatography jar with tops ground to close tolerances to insure a tight cover fit. Covers are not supplied.
These cylindrical chromatography jars are made of Pyrex® borosilicate glass.
The jar edge is flat within 0.102 mm (0.004") on 6.6 L (1.7 gal.) jar, all others are flat within 0.254 mm (0.01").
Caution: Note: Do not use with heat, vacuum, or pressure applications.