VWR® Talon® Stainless Steel Fume Hood Lab-Frame Kits
Supplier: VWR International
14224-400 14224-402 14224-404 14224-406
14224-400EA 2465.5 CAD
14224-400 14224-402 14224-404 14224-406
VWR® Talon® Stainless Steel Fume Hood Lab-Frame Kits
Retort Rods
Choose from four kits specifically designed to fit within laboratory fume hoods.
  • Stainless steel construction of rods
  • Nickel-plated zinc connectors and lab frame feet
  • Variety of kits designed to fit within laboratory fume hoods
The 1.5m (5') kit includes four 127 cm (50") horizontal rods, five 91.4 cm (36") vertical rods, two 30.5 cm (12") base rods, two 45.7 cm (18") rear support rods, eight 5.1 cm (2") rods, 34 lattice connectors, four rod-end connectors, and four Lab Frame feet.
The 1.2m (4') kit includes four 96.5 cm (38") horizontal rods, fourMore Product Information