BRAND® Disposable Plastic Cuvettes, Acrylic and Polystyrene, BrandTech
Supplier: Brandtech
759070D 759075D 759080D 759081D 759085D 759086D 759076D 759071D
47743-940PK 317.35 CAD
47743-940 47744-642 47744-646 47744-648 47744-650 47744-652 47744-644 47751-090
BRAND® Disposable Plastic Cuvettes, Acrylic and Polystyrene, BrandTech
Cuvettes Spectrophotometer Cuvettes
Disposable cuvettes are constructed of high-quality polymethyl methacrylate (PMMA or "acrylic") or polystyrene (PS), and are excellent for most spectrophotometry applications in the visible range
  • 10 mm disposable cuvettes with two optical sides
  • Fits most common spectrophotometers
  • Mold-cavity match for minimum variability
Acrylic cuvettes are suitable for assays between 300nm and 900nm. Choose polystyrene cuvettes for routine assays between 340nm and 900nm.
Cuvettes include an arrow mark to indicate direction of transmission and reduce variation. They are grouped by manufacturing mold cavity number to ensure the lowest variation in extinction coefficient, and are packaged in low dust, low scratch material.
They provide accurate, reliable results and are available in macro and semi-macro sizes to accommodate most sample volumes. Cuvettes are compatible with most spectrophotometers and photometers.
Ordering information: For disposible cuvettes for wavelengths below 300nm, see 47743-834 series.