Cuvettes Plus™ Electroporation Cuvettes, BTX™
Supplier: Harvard Apparatus
45-0124 450125 450126
CA58017-890PK 677.73 CAD
CA58017-890 CA58017-895 CA58017-900
Cuvettes Plus™ Electroporation Cuvettes, BTX™
Cuvettes Electroporation Cuvettes
Designed for use in the electroporation of bacteria, yeast, mammalian, and plant cells.
The cuvettes are molded with polished aluminum electrodes and medical grade polycarbonate, washed prior to packaging, and sterilized using gamma irradiation. Gap sizes of 1mm, 2mm, and 4mm are available for high field strengths up to 25.0kV/cm. Each cuvette has a color-coded, leak-resistant round cap that allows single-handed manipulation. Simply discard cuvettes carefully after use. Cuvettes fit all BTX™ and most comparable electroporators, including those from Bio-Rad®, Eppendorf®, and Invitrogen®.
More Product Information