MPC Series Plastic Couplings, Colder Products Company
Supplier: CPC (Colder Products Company)
MPC22004T03M MPC17006T39 MPC17004T03 MPC22004T39M MPC17006T03 MPC17004T39 MPC22006T39M MPC22006T03M
16001-588PK 2036.16 CAD
16001-588 16001-600 16001-596 16001-586 16001-598 16001-592 16001-594 16001-590
MPC Series Plastic Couplings, Colder Products Company
Fittings and Connectors
Couplings allow rapid disconnect and reconnect with one hand and without the need for tools
Couplings are available in polysulfone (amber tint) or polycarbonate (purple tint) construction. Coupling inserts include seals constructed of clear silicone. All products meet USP Class VI criteria.
They feature an ergonomic thumb latch and an audible click when connected, assuring the user of a secure connection. Couplings can be rotated when connected, reducing the inconvenience of tube kinks. These items are sterilizable and reusable, yet economical enough to allow disposability.
Certifications: All products meet USP Class VI criteria and feature BSE/TSE-free materials (ADCF).